Titanium powder, -325 mesh, 99% (metals basis)
Titanium powder, -325 mesh, 99% (metals basis)
Titanium powder, -325 mesh, 99% (metals basis)
Titanium powder, -325 mesh, 99% (metals basis)
Thermo Scientific Chemicals
Titanium powder, -325 mesh, 99% (metals basis)
CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF010386.D32 x 1 kg
Catalog number ALF010386.D3
Price (MYR)
5,591.00
EA
Add to cart
Quantity:
2 x 1 kg
Request bulk or custom format
Price (MYR)
5,591.00
EA
Add to cart
Specifications
Chemical Name or MaterialTitanium
ColorGray
CAS7440-32-6
Health Hazard 1H250-H500
Health Hazard 3P210-P222-P280-P335+P334-P370+P378q