Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis)
Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis)
Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis)
Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis)
Thermo Scientific Chemicals
Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis)
Form: approx. 11.3g/100x100mm | CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 500 mm
100 x 100 mm
Catalog number ALF010385.HP
Price (MYR)
1,441.00
EA
Add to cart
Quantity:
100 x 500 mm
Request bulk or custom format
Price (MYR)
1,441.00
EA
Add to cart
Specifications
Chemical Name or MaterialTitanium
ColorSilver to Gray
Name NoteAnnealed
CAS7440-32-6
AppearanceSilver Gray foil