Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals
Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Form: Approx. 10.4g/25 x 25mm | CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Catalog NumberQuantity
ALF010356.FI50 x 50 mm
Catalog number ALF010356.FI
Price (MYR)
2,225.00
EA
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (MYR)
2,225.00
EA
Add to cart
Specifications
Chemical Name or MaterialTantalum
ColorGray to Silver
CAS7440-25-7
AppearanceGray to silver foil