Sodium, dispersion, 40% in oil
Sodium, dispersion, 40% in oil
Sodium, dispersion, 40% in oil
Sodium, dispersion, 40% in oil
Thermo Scientific Chemicals
Sodium, dispersion, 40% in oil
CAS: 7440-23-5 | Na | 22.99 g/mol
Have Questions?
Catalog NumberQuantity
ALF010340.30250 g
Catalog number ALF010340.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium
ColorWhite
CAS7440-23-5
Health Hazard 1H260-H314-H335
Health Hazard 3P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c