Rhenium foil, 0.25mm (0.01in) thick, 99.97% (metals basis)
Rhenium foil, 0.25mm (0.01in) thick, 99.97% (metals basis)
Rhenium foil, 0.25mm (0.01in) thick, 99.97% (metals basis)
Rhenium foil, 0.25mm (0.01in) thick, 99.97% (metals basis)
Thermo Scientific Chemicals
Rhenium foil, 0.25mm (0.01in) thick, 99.97% (metals basis)
Form: Approx. 3.29g/25 x 25mm; Solid in various forms | CAS: 7440-15-5 | Re | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number ALF010309.FI
Price (MYR)
8,004.00
EA
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (MYR)
8,004.00
EA
Add to cart
Specifications
Chemical Name or MaterialRhenium
ColorSilver to Gray
CAS7440-15-5
AppearanceSilver gray foil