Platinum wire, 0.025mm (0.001in) dia, 99.95% (metals basis)
Platinum wire, 0.025mm (0.001in) dia, 99.95% (metals basis)
Platinum wire, 0.025mm (0.001in) dia, 99.95% (metals basis)
Platinum wire, 0.025mm (0.001in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals
Platinum wire, 0.025mm (0.001in) dia, 99.95% (metals basis)
Form: Approx.0.01g/m | CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF010292.G55 m
Catalog number ALF010292.G5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 m
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum
ColorWhite
CAS7440-06-4
AppearanceSilvery white wire