Platinum wire, 0.5mm (0.02in) dia, hard, 99.95% (metals basis)
Platinum wire, 0.5mm (0.02in) dia, hard, 99.95% (metals basis)
Platinum wire, 0.5mm (0.02in) dia, hard, 99.95% (metals basis)
Platinum wire, 0.5mm (0.02in) dia, hard, 99.95% (metals basis)
Platinum wire, 0.5mm (0.02in) dia, hard, 99.95% (metals basis)
Thermo Scientific Chemicals
Platinum wire, 0.5mm (0.02in) dia, hard, 99.95% (metals basis)
Form: Approx.4.21g/m; Temper: as drawn | CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 cm
25 cm
Catalog number ALF010286.BY
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum
ColorWhite
Name NoteHard
CAS7440-06-4
AppearanceSilvery white wire