Niobium powder, -325 mesh, 99.8% (metals basis)
Niobium powder, -325 mesh, 99.8% (metals basis)
Niobium powder, -325 mesh, 99.8% (metals basis)
Niobium powder, -325 mesh, 99.8% (metals basis)
Thermo Scientific Chemicals
Niobium powder, -325 mesh, 99.8% (metals basis)
CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Catalog NumberQuantity
ALF010275.22100 g
Catalog number ALF010275.22
Price (MYR)
1,101.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,101.00
EA
Add to cart
Specifications
Chemical Name or MaterialNiobium powder
CAS7440-03-1
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228
Flammable solid.
Health Hazard 3P210-P240-P241-P280-P370+P378q