Manganese powder, -325 mesh, 99.3% (metals basis)
Manganese powder, -325 mesh, 99.3% (metals basis)
Manganese powder, -325 mesh, 99.3% (metals basis)
Manganese powder, -325 mesh, 99.3% (metals basis)
Thermo Scientific Chemicals
Manganese powder, -325 mesh, 99.3% (metals basis)
Form: Apparent density 2.6-3.5g/cm3 | CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Catalog NumberQuantity
ALF010238.30250 g
Catalog number ALF010238.30
Price (MYR)
445.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
445.00
EA
Add to cart
Specifications
Chemical Name or MaterialManganese
ColorGray
CAS7439-96-5
Health Hazard 1H228-H319
AppearanceGray powder