Manganese pieces, irregular, 99.9% (metals basis)
Manganese pieces, irregular, 99.9% (metals basis)
Manganese pieces, irregular, 99.9% (metals basis)
Manganese pieces, irregular, 99.9% (metals basis)
Thermo Scientific Chemicals
Manganese pieces, irregular, 99.9% (metals basis)
CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Catalog NumberQuantity
ALF010236.30250 g
Catalog number ALF010236.30
Price (MYR)
424.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
424.00
EA
Add to cart
Specifications
Chemical Name or MaterialManganese
ColorGray
CAS7439-96-5
AppearanceGray irregularly shaped pieces
Density7.3g/cm3 at 20°C