Lead powder, -200 mesh, 99.9% (metals basis)
Lead powder, -200 mesh, 99.9% (metals basis)
Lead powder, -200 mesh, 99.9% (metals basis)
Lead powder, -200 mesh, 99.9% (metals basis)
Lead powder, -200 mesh, 99.9% (metals basis)
Thermo Scientific Chemicals
Lead powder, -200 mesh, 99.9% (metals basis)
CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Catalog NumberQuantity
ALF010223.30250 g
Catalog number ALF010223.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLead
ColorBlack to Gray
CAS7439-92-1
Health Hazard 1H350-H360FD-H362-H372
Health Hazard 3P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c