Gold wire, 0.2mm (0.008in) dia, 99.9% (metals basis)
Gold wire, 0.2mm (0.008in) dia, 99.9% (metals basis)
Gold wire, 0.2mm (0.008in) dia, 99.9% (metals basis)
Gold wire, 0.2mm (0.008in) dia, 99.9% (metals basis)
Thermo Scientific Chemicals
Gold wire, 0.2mm (0.008in) dia, 99.9% (metals basis)
Form: Approx.0.62g/m | CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
Catalog number ALF010195.G5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 m
Request bulk or custom format
Specifications
Chemical Name or MaterialGold
ColorGolden
CAS7440-57-5
AppearanceGold colored wire