Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
Thermo Scientific Chemicals
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF010191.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGermanium pieces
Name Note3 to 9mm (0.1 to 0.4 in.)
CAS7440-56-4
Recommended StorageAmbient temperatures
EINECS Number231-164-3