Chromium powder, -325 mesh, 99% (metals basis)
Chromium powder, -325 mesh, 99% (metals basis)
Chromium powder, -325 mesh, 99% (metals basis)
Chromium powder, -325 mesh, 99% (metals basis)
Thermo Scientific Chemicals
Chromium powder, -325 mesh, 99% (metals basis)
CAS: 7440-47-3 | Cr | 52.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF010148.36
Price (MYR)
1,649.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,649.00
EA
Add to cart
Specifications
Chemical Name or MaterialChromium
ColorGray
CAS7440-47-3
Health Hazard 1H228
AppearanceGray powder