Cesium, 99.98% (metals basis)
Cesium, 99.98% (metals basis)
Cesium, 99.98% (metals basis)
Cesium, 99.98% (metals basis)
Thermo Scientific Chemicals
Cesium, 99.98% (metals basis)
CAS: 7440-46-2 | Cs | 132.91 g/mol
Have Questions?
Catalog NumberQuantity
ALF010146.1425 g
Catalog number ALF010146.14
Price (MYR)
5,585.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
5,585.00
EA
Add to cart
Specifications
Chemical Name or MaterialCesium
Name Note(metals basis)
CAS7440-46-2
Health Hazard 1H260-H314-H361
Health Hazard 2GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.