Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%
Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr &mid;&lt;0.8%
Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr &mid;&lt;0.8%
Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr &mid;&lt;0.8%
Thermo Scientific Chemicals
Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%
Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps | CAS: 7440-39-3 | Ba | 137.33 g/mol
Have Questions?
Catalog NumberQuantity
ALF010103.22100 g
Catalog number ALF010103.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBarium rod
ColorGray
Name Note22mm (0.9 in.) dia.
CAS7440-39-3
Health Hazard 1H260-H304
View more
Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only