Antimony powder, -200 mesh, 99.5% (metals basis)
Antimony powder, -200 mesh, 99.5% (metals basis)
Antimony powder, -200 mesh, 99.5% (metals basis)
Antimony powder, -200 mesh, 99.5% (metals basis)
Thermo Scientific Chemicals
Antimony powder, -200 mesh, 99.5% (metals basis)
CAS: 7440-36-0 | Sb | 121.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF010099.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAntimony powder
ColorGray
CAS7440-36-0
Health Hazard 1H335-H351-H373
Health Hazard 2GHS H Statement
H301-H332