Molybdenum wire, 1.0mm (0.04in) dia, annealed, 99.94% (metals basis)
Molybdenum wire, 1.0mm (0.04in) dia, annealed, 99.94% (metals basis)
Molybdenum wire, 1.0mm (0.04in) dia, annealed, 99.94% (metals basis)
Molybdenum wire, 1.0mm (0.04in) dia, annealed, 99.94% (metals basis)
Thermo Scientific Chemicals
Molybdenum wire, 1.0mm (0.04in) dia, annealed, 99.94% (metals basis)
Form: approx. 8.0g/m, Temper: soft | CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Catalog NumberQuantity
ALF010039.G925 m
Catalog number ALF010039.G9
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 m
Request bulk or custom format
Specifications
Chemical Name or MaterialMolybdenum
ColorGray
CAS7439-98-7
AppearanceDark gray wire