Titanium foil, 0.127mm (0.005in) thick, annealed, 99% (metals basis)
Titanium foil, 0.127mm (0.005in) thick, annealed, 99% (metals basis)
Titanium foil, 0.127mm (0.005in) thick, annealed, 99% (metals basis)
Titanium foil, 0.127mm (0.005in) thick, annealed, 99% (metals basis)
Thermo Scientific Chemicals
Titanium foil, 0.127mm (0.005in) thick, annealed, 99% (metals basis)
CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
150 x 150 mm
50 x 50 mm
Catalog number ALF000971.GT
Price (MYR)
986.00
EA
Add to cart
Quantity:
150 x 150 mm
Request bulk or custom format
Price (MYR)
986.00
EA
Add to cart
Specifications
Chemical Name or MaterialTitanium foil
CAS7440-32-6
Recommended StorageAmbient temperatures
Quantity150 x 150 mm
EINECS Number231-142-3