Gold powder, spherical, <10 micron, 99.95% (metals basis)
Gold powder, spherical, &lt;10 micron, 99.95% (metals basis)
Gold powder, spherical, &lt;10 micron, 99.95% (metals basis)
Gold powder, spherical, &lt;10 micron, 99.95% (metals basis)
Thermo Scientific Chemicals
Gold powder, spherical, <10 micron, 99.95% (metals basis)
CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Catalog NumberQuantity
ALF000765.031 g
Catalog number ALF000765.03
Price (MYR)
1,836.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,836.00
EA
Add to cart
Specifications
Chemical Name or MaterialGold powder
CAS7440-57-5
Recommended StorageAmbient temperatures
Quantity1 g
EINECS Number231-165-9