Iron powder, -200 mesh, 99+% (metals basis), Thermo Scientific Chemicals
Iron powder, -200 mesh, 99+% (metals basis), Thermo Scientific Chemicals
Iron powder, -200 mesh, 99+% (metals basis), Thermo Scientific Chemicals
Iron powder, -200 mesh, 99+% (metals basis), Thermo Scientific Chemicals
Iron powder, -200 mesh, 99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iron powder, -200 mesh, 99+% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF000737.30250 g
Catalog number ALF000737.30
Price (MYR)
435.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
435.00
EA
Add to cart
Specifications
Chemical Name or MaterialIron powder
CAS7439-89-6
Health Hazard 1H500-X
Health Hazard 2GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P235+P410-P280