Silver powder, -100 mesh, 99.95% (metals basis)
Silver powder, -100 mesh, 99.95% (metals basis)
Silver powder, -100 mesh, 99.95% (metals basis)
Silver powder, -100 mesh, 99.95% (metals basis)
Thermo Scientific Chemicals
Silver powder, -100 mesh, 99.95% (metals basis)
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF000690.22
Price (MYR)
2,981.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,981.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
CAS7440-22-4
Health Hazard 1H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
AppearanceSilver-colored powder