Yttrium pieces, 99.9% (REO)
Yttrium pieces, 99.9% (REO)
Yttrium pieces, 99.9% (REO)
Yttrium pieces, 99.9% (REO)
Thermo Scientific Chemicals
Yttrium pieces, 99.9% (REO)
CAS: 7440-65-5 | Y | 88.91 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALF000615.0910 gUnder Argon
Catalog number ALF000615.09
Price (MYR)
986.00
EA
Add to cart
Quantity:
10 g
Packaging:
Under Argon
Request bulk or custom format
Price (MYR)
986.00
EA
Add to cart
Specifications
Chemical Name or MaterialYttrium
Name NoteRare Earth Oxides Content
CAS7440-65-5
AppearanceGray pieces
Density4.47g/cm3