Nickel pieces, 99.9+% (metals basis)
Nickel pieces, 99.9+% (metals basis)
Nickel pieces, 99.9+% (metals basis)
Nickel pieces, 99.9+% (metals basis)
Thermo Scientific Chemicals
Nickel pieces, 99.9+% (metals basis)
CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Catalog NumberQuantity
ALF000434.22100 g
Catalog number ALF000434.22
Price (MYR)
417.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
417.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel pieces
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c