Titanium foil, 0.025mm (0.001in) thick, 99.94% (metals basis)
Titanium foil, 0.025mm (0.001in) thick, 99.94% (metals basis)
Titanium foil, 0.025mm (0.001in) thick, 99.94% (metals basis)
Titanium foil, 0.025mm (0.001in) thick, 99.94% (metals basis)
Thermo Scientific Chemicals
Titanium foil, 0.025mm (0.001in) thick, 99.94% (metals basis)
Form: Approx. 0.2g/25 x 75mm | CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF000360.FH25 x 75 mm
Catalog number ALF000360.FH
Price (MYR)
1,019.00
EA
Add to cart
Quantity:
25 x 75 mm
Request bulk or custom format
Price (MYR)
1,019.00
EA
Add to cart
Specifications
Chemical Name or MaterialTitanium
ColorSilver to Gray
CAS7440-32-6
AppearanceSilver Gray foil
Density4.506g/cm3