Platinum foil, 0.127mm (0.005in) thick, 99.9% (metals basis)
Platinum foil, 0.127mm (0.005in) thick, 99.9% (metals basis)
Platinum foil, 0.127mm (0.005in) thick, 99.9% (metals basis)
Platinum foil, 0.127mm (0.005in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals
Platinum foil, 0.127mm (0.005in) thick, 99.9% (metals basis)
Form: Approx.1.7g/25 x 25mm | CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF000261.FF25 x 25 mm
Catalog number ALF000261.FF
Price (MYR)
3,442.00
EA
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (MYR)
3,442.00
EA
Add to cart
Specifications
Chemical Name or MaterialPlatinum
ColorWhite
CAS7440-06-4
AppearanceSilvery white foil