Phosphorus lump, red, Puratronic™, 99.999+% (metals basis)
Phosphorus lump, red, Puratronic™, 99.999+% (metals basis)
Phosphorus lump, red, Puratronic™, 99.999+% (metals basis)
Phosphorus lump, red, Puratronic™, 99.999+% (metals basis)
Thermo Scientific Chemicals
Phosphorus lump, red, Puratronic™, 99.999+% (metals basis)
CAS: 7723-14-0 | P | 30.97 g/mol
Have Questions?
Catalog NumberQuantity
ALF000248.1850 g
Catalog number ALF000248.18
Price (MYR)
4,439.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
4,439.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhosphorus lump, red
CAS7723-14-0
ColorRed to Brown
Health Hazard 1Danger
Health Hazard 2Hazard statements
H228 Flammable solid