Nickel foil, 0.025mm (0.001in) thick, annealed, 99.5% (metals basis)
Nickel foil, 0.025mm (0.001in) thick, annealed, 99.5% (metals basis)
Nickel foil, 0.025mm (0.001in) thick, annealed, 99.5% (metals basis)
Nickel foil, 0.025mm (0.001in) thick, annealed, 99.5% (metals basis)
Thermo Scientific Chemicals
Nickel foil, 0.025mm (0.001in) thick, annealed, 99.5% (metals basis)
Form: approx. 1.4g/25x250mm | CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Catalog NumberQuantity
ALF000228.FW25 x 500 mm
Catalog number ALF000228.FW
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 x 500 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel
ColorSilver to Gray
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 3P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c