Copper foil, 0.127mm (0.005in) thick, annealed, 99.99+% (metals basis)
Copper foil, 0.127mm (0.005in) thick, annealed, 99.99+% (metals basis)
Copper foil, 0.127mm (0.005in) thick, annealed, 99.99+% (metals basis)
Copper foil, 0.127mm (0.005in) thick, annealed, 99.99+% (metals basis)
Thermo Scientific Chemicals
Copper foil, 0.127mm (0.005in) thick, annealed, 99.99+% (metals basis)
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
ALF000095.FI50 x 50 mm
Catalog number ALF000095.FI
Price (MYR)
745.00
EA
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (MYR)
745.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper foil
Name Note0.127mm (0.005 in.) thick, Annealed
CAS7440-50-8
Recommended StorageAmbient temperatures
Quantity50 x 50 mm