Titanium foil, 0.74mm (0.029in) thick, annealed, 99.4+% (metals basis)
Titanium foil, 0.74mm (0.029in) thick, annealed, 99.4+% (metals basis)
Titanium foil, 0.74mm (0.029in) thick, annealed, 99.4+% (metals basis)
Titanium foil, 0.74mm (0.029in) thick, annealed, 99.4+% (metals basis)
Thermo Scientific Chemicals
Titanium foil, 0.74mm (0.029in) thick, annealed, 99.4+% (metals basis)
CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF000029.GX200 x 300 mm
Catalog number ALF000029.GX
Price (MYR)
1,019.00
EA
Add to cart
Quantity:
200 x 300 mm
Request bulk or custom format
Price (MYR)
1,019.00
EA
Add to cart
Specifications
Chemical Name or MaterialTitanium foil
ColorGray
Name Note0.74mm (0.029 in.) thick, Annealed
CAS7440-32-6