Check Order Status
2,5-dichlorophenyl isothiocyanate, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific
2,5-dichlorophenyl isothiocyanate, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
ACRSB01698EB
25 g
Catalog number
ACRSB01698EB
Price (MYR)
792.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
792.00
EA
Add to cart
Specifications
Chemical Name or Material
2, 5-Dichlorophenyl isothiocyanate
CAS
3386-42-3
Health Hazard 1
GHS Signal Word:
Danger
Assay Percent Range
97+%
Molecular Formula
C7H3Cl2NS
View more