Methyl heptadecanoate, Thermo Scientific™
Product Image
Methyl heptadecanoate, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ACR4693000101 g
Catalog number ACR469300010
Price (MYR)
577.00
EA
Add to cart
Quantity:
1 g
Price (MYR)
577.00
EA
Add to cart
Specifications
Alpha VectorMETHYLHEPTADECANOATE
Chemical Name or MaterialMethyl heptadecanoate
Shelf Life5 years
CAS1731-92-6
Melting Point29°C to 30°C
ColorColorless or White to Yellow
Recommended StorageNormal conditions
EINECS Number217-055-3
Boiling Point152°C to 153°C
Flash Point>110°C
Infrared SpectrumConforms
PackagingGlass bottle
Molecular FormulaC18 H36 O2
MDL Number00009001
Quantity1 g
SynonymMethyl margarate
Molecular Weight (g/mol)284.48
Physical FormLiquid or Powder
Unit SizeEA