Oxibendazole, Thermo Scientific™
Oxibendazole, Thermo Scientific™
Thermo Scientific Chemicals
Oxibendazole, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ACR4688100101 g
Catalog number ACR468810010
Price (MYR)
803.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
803.00
EA
Add to cart
Specifications
Chemical Name or MaterialOxibendazole
PackagingGlass Bottle
CAS20559-55-1
Certificate of AnalysisAvailable
Recommended StorageNormal conditions
View more