Myclobutanil
Thermo Scientific Chemicals
Myclobutanil
CAS: 88671-89-0 | C15H17ClN4 | 288.78 g/mol
Have Questions?
Catalog NumberQuantity
ACR4671400101 g
Catalog number ACR467140010
Price (MYR)
556.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
556.00
EA
Add to cart
Specifications
Chemical Name or MaterialMyclobutanil
CAS88671-89-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Harmful if swallowed.
Causes serious eye irritation.
Suspected of damaging the unborn child.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.