Dinotefuran
Thermo Scientific Chemicals
Dinotefuran
CAS: 165252-70-0 | C7H14N4O3, C7H14N4O3 | 202.21 g/mol
Have Questions?
Catalog NumberQuantity
ACR467092500250 mg
Catalog number ACR467092500
Price (MYR)
614.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (MYR)
614.00
EA
Add to cart
Specifications
Chemical Name or MaterialDinotefuran
CAS165252-70-0
Quantity250 mg
SolubilitySolubility aq. soln.: 39.8 g/L (20°C). Other solubilities: soluble in ethanol, methanol and dimethyl sulfoxid, e
Melting Point106.0°C to 111.0°C