Maltodextrin 12
Thermo Scientific Chemicals
Maltodextrin 12
CAS: 9050-36-6 | C6H12O6 | 180.16 g/mol
Have Questions?
Catalog NumberQuantity
ACR46668050050 g
Catalog number ACR466680500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMaltodextrin 12
CAS9050-36-6
Melting Point240.0°C
EINECS Number232-940-4
Molecular FormulaC6H12O6