Raloxifene hydrochloride
Raloxifene hydrochloride
Thermo Scientific Chemicals
Raloxifene hydrochloride
CAS: 82640-04-8 | C28H28ClNO4S, C28H27NO4S·HCl | 510.05 g/mol
Have Questions?
Catalog NumberQuantity
ACR4665000101 g
Catalog number ACR466500010
Price (MYR)
1,994.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,994.00
EA
Add to cart
Specifications
Chemical Name or MaterialRaloxifene hydrochloride
CAS82640-04-8
Quantity1 g
SolubilitySolubility aq. soln.: .. Other solubilities: soluble in dmso
Melting Point263.0°C