Bambuterol hydrochloride, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals
Bambuterol hydrochloride, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR4649100505 g
Catalog number ACR464910050
Price (MYR)
2,604.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
2,604.00
EA
Add to cart
Specifications
Chemical Name or MaterialBambuterol hydrochloride
CAS81732-46-9
Melting Point225.0°C to 229.0°C
Molecular FormulaC18H30ClN3O5
MDL NumberMFCD03427293