Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
CAS: 125494-59-9 | C17H29Cl2NO | 334.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR462200010
Price (MYR)
0.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
0.00
EA
Add to cart
Specifications
Chemical Name or MaterialSibutramine hydrochloride monohydrate
CAS125494-59-9
Molecular FormulaC17H29Cl2NO
Quantity1 g
InChI KeyKFNNPQDSPLWLCX-UHFFFAOYNA-N