Ammonium hydroxide, for HPLC, 35% solution in water
Ammonium hydroxide, for HPLC, 35% solution in water
Ammonium hydroxide, for HPLC, 35% solution in water
Ammonium hydroxide, for HPLC, 35% solution in water
Thermo Scientific Chemicals
Ammonium hydroxide, for HPLC, 35% solution in water
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Catalog NumberQuantity
ACR460801000100 mL
Catalog number ACR460801000
Price (MYR)
495.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
495.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium hydroxide
CAS1336-21-6
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Very toxic to aquatic life with long lasting effects.
CAS7732-18-5