Ertapenem sodium, 85%
Ertapenem sodium, 85%
Ertapenem sodium, 85%
Ertapenem sodium, 85%
Ertapenem sodium, 85%
Have Questions?
Catalog NumberQuantity
ACR460642500250 mg
Catalog number ACR460642500
Price (MYR)
824.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (MYR)
824.00
EA
Add to cart
Specifications
Chemical Name or MaterialErtapenem sodium
CAS153773-82-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air