Meloxicam, 99-101%
Meloxicam, 99-101%
Meloxicam, 99-101%
Thermo Scientific Chemicals
Meloxicam, 99-101%
CAS: 71125-38-7 | C14H13N3O4S2 | 351.40 g/mol
Have Questions?
Catalog NumberQuantity
ACR45955025025 g
Catalog number ACR459550250
Price (MYR)
1,627.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,627.00
EA
Add to cart
Specifications
Chemical Name or MaterialMeloxicam
CAS71125-38-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent