Celecoxib, 98%, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals
Celecoxib, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR456451000100 mg
Catalog number ACR456451000
Price (MYR)
1,441.00
EA
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (MYR)
1,441.00
EA
Add to cart
Specifications
Chemical Name or MaterialCelecoxib
CAS169590-42-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement May damage the unborn child.
Health Hazard 3GHS P Statement Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.