Montmorillonite K-10, granules
Montmorillonite K-10, granules
Montmorillonite K-10, granules
Montmorillonite K-10, granules
Acros Organics
Montmorillonite K-10, granules
CAS: 1318-93-0 | Al2H2O12Si4 | 360.307 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Catalog number ACR456071000
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMontmorillonite K-10
CAS1318-93-0
Quantity100 g
EINECS Number215-288-5
Molecular FormulaAl2H2O12Si4