Nystatin, 85+%
Nystatin, 85+%
Nystatin, 85+%
Thermo Scientific Chemicals
Nystatin, 85+%
Nystatin, CAS # 1400-61-9, is a polyene compound that shows anti-bacterial and antifungal activities. It is also an ionophore that increases the membrane permeability to some ions.
Have Questions?
Catalog NumberQuantity
ACR4555000101 g
Catalog number ACR455500010
Price (MYR)
496.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
496.00
EA
Add to cart
Specifications
Chemical Name or MaterialNystatin
TypeNystatin
Physical FormPowder
CAS1400-61-9
Melting Point>160.0°C
View more
General Description
• Nystatin is a polyene compound that shows anti-bacterial and antifungal activities against Streptomyces noursei, Streptomyces aureus, and other Streptomyces species
• It is also an ionophore with the capacity to increase membrane permeability
• Nystatin is a channel-forming ionophore and exerts its activity via formation of a membrane-spanning pore in the fungal plasma membrane, thus altering membrane permeability, allowing the leakage of intracellular contents, and the subsequent disruption of electrochemical gradients essential for proper cell function
Applications
• In in vitro experiments, Nystatin is an antifungal that is both fungistatic and fungicidal against a wide variety of yeasts and yeast-like fungi
• It has a higher selectivity for fungal cells over mammalian cells since nystatin presents greater binding affinity with ergosterol, a key sterol found in fungal cell walls, as opposed to its mammalian counterpart, cholesterol
• This compound is not active against organisms that do not contain sterols in their cell membrane (for example, bacteria)
• It is used on human macrophage cell line U937, to study its effect on the production of interleukin-8 by silver nanoparticle-treated U937 cell line
• It has the ability to prevent cell culture contamination as well as induce interleukin-1 and -8, and tumor necrosis factor α secretion in TLR2-expressing THP1 cells
• It modulates lipid raft profile in mice macrophages
RUO – Research Use Only
General References:
  1. R Holz.; A Finkelstein. The water and nonelectrolyte permeability induced in thin lipid membranes by the polyene antibiotics nystatin and amphotericin B. The Journal of general physiology. 1970, 56, (1), 125-145.
  2. Kim, S.; Choi, I.H. Phagocytosis and endocytosis of silver nanoparticles induce interleukin-8 production in human macrophages. Yonsei Med J. 2012, 53, (3), 654-657.
  3. Razonable, R.R.; Henault, M., Watson, H.L.; Paya, C.V. Nystatin induces secretion of interleukin (IL)-1beta, IL-8, and tumor necrosis factor alpha by a toll-like receptor-dependent mechanism. Antimicrob Agents Chemother. 2005, 49, (8), 3546-3549.