Clarithromycin, 97%
Clarithromycin, 97%
Clarithromycin, 97%
Thermo Scientific Chemicals
Clarithromycin, 97%
CAS: 81103-11-9 | C38H69NO13 | 747.95 g/mol
Have Questions?
Catalog NumberQuantity
ACR4552200101 g
Catalog number ACR455220010
Price (MYR)
1,582.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,582.00
EA
Add to cart
Specifications
Chemical Name or MaterialClarithromycin
CAS81103-11-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.