Amoxicillin, 96%
Amoxicillin, 96%
Amoxicillin, 96%
Thermo Scientific Chemicals
Amoxicillin, 96%
CAS: 26787-78-0 | C16H19N3O5S | 365.404 g/mol
Have Questions?
Catalog NumberQuantity
ACR45514025025 g
Catalog number ACR455140250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmoxicillin
TypeAmoxicillin
Physical FormSolid
CAS26787-78-0
Health Hazard 1Danger