Graphene, nanoplatelets 2-10 nm
Graphene, nanoplatelets 2-10 nm
Graphene, nanoplatelets 2-10 nm
Thermo Scientific Chemicals
Graphene, nanoplatelets 2-10 nm
CAS: 1034343-98-0 | Cx | 16.04 g/mol
Have Questions?
Catalog NumberQuantity
ACR45316050050 g
Catalog number ACR453160500
Price (MYR)
1,750.00
Each
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,750.00
Each
Specifications
Chemical Name or MaterialGraphene
CAS1034343-98-0
Melting Point3650.0°C
Molecular FormulaCx
MDL NumberMFCD00144065