Perchloric acid, extra pure, ca. 70% solution in water
Perchloric acid, extra pure, ca. 70% solution in water
Perchloric acid, extra pure, ca. 70% solution in water
Perchloric acid, extra pure, ca. 70% solution in water
Thermo Scientific Chemicals
Perchloric acid, extra pure, ca. 70% solution in water
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR452850010
Price (MYR)
1,129.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,129.00
EA
Add to cart
Specifications
Chemical Name or MaterialPerchloric Acid
CAS7601-90-3
Melting Point-18.0°C
Boiling Point203.0°C
Density1.6600g/mL