Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Thermo Scientific Chemicals
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
ACR4507200252.5 L
Catalog number ACR450720025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS7647-01-0, 67-63-0
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Harmful if inhaled. Highly flammable liquid and vapor.
CAS67-63-0